Novus Biologicals
Manufacturer Code:NBP132823
Catalog # NBP132823
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human UAP1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGX; AGX AGX1 AGX2 Antigen X EC 2.7.7 EC 2.7.7.23 EC 2.7.7.27 SPAG2 sperm associated antigen 2 Sperm-associated antigen 2 UDP-N-acetylhexosamine pyrophosphorylase UDP-N-acteylglucosamine pyrophosphorylase 1; AGX-1; AGX-2; Antigen X; sperm associated antigen 2; Sperm-associated antigen 2; testis tissue sperm-binding protein Li 37a; UDP-N-acetylgalactosamine pyrophosphorylase; UDP-N-acetylglucosamine diphosphorylase 1; UDP-N-acetylglucosamine pyrophosphorylase; UDP-N-acetylhexosamine pyrophosphorylase; UDP-N-acetylhexosamine pyrophosphorylase 1
Gene Aliases: AGX; AGX1; AGX2; SPAG2; UAP1
UniProt ID: (Human) Q16222
Entrez Gene ID: (Human) 6675
Molecular Function:
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.