Novus Biologicals
Manufacturer Code:NBP236444AF405
Catalog # NBP236444W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | AE-5 |
Immunogen | Nuclei of myeloid leukemia biopsy cells |
Conjugate | Alexa Fluor 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: FLJ10128 KIAA1561MGC141969 MGC141967 nuclear membrane binding protein NUCLING uveal autoantigen with coiled coil domains and ankyrin repeats uveal autoantigen with coiled-coil domains and ankyrin repeats; nuclear membrane binding protein; uveal autoantigen with coiled coil domains and ankyrin repeats; Uveal autoantigen with coiled-coil domains and ankyrin repeats
Gene Aliases: KIAA1561; NUCLING; UACA
UniProt ID: (Human) Q9BZF9
Entrez Gene ID: (Human) 55075
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.