Invitrogen
Manufacturer Code:PA530442
Catalog # PIPA530442
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 252 and 426 of Human U2AF65 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hU2AF(65); hU2AF65; Splicing factor U2AF 65 kDa subunit; U2 (RNU2) small nuclear RNA auxiliary factor 2; U2 auxiliary factor 65 kDa subunit; U2 small nuclear ribonucleoprotein auxiliary factor (65kD); U2 small nuclear ribonucleoprotein auxiliary factor (U2AF), 65 kDa; U2 snRNP auxiliary factor large subunit; U2AF65 U2 (RNU2) small nuclear RNA auxiliary factor 2 U2 auxiliary factor 65 kDa subunit U2 small nuclear ribonucleoprotein auxiliary factor (65kD) U2 snRNP auxiliary factor large subunit hU2AF(65) hU2AF65 splicing factor U2AF 65 kD subunit splicing factor U2AF
Gene Aliases: 65kDa; U2AF2; U2AF65
UniProt ID: (Human) P26368, (Mouse) P26369
Entrez Gene ID: (Human) 11338, (Mouse) 22185
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.