Invitrogen
Manufacturer Code:PA518826
Catalog # PIPA518826
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (EDGERAVAELSNR) corresponding to the internal amino acids of U2AF1L4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Splicing factor U2AF 26 kDa subunit; U2 auxiliary factor 26; U2 small nuclear RNA auxiliary factor 1-like 3; U2 small nuclear RNA auxiliary factor 1-like 4; U2 small nuclear RNA auxiliary factor 1-like protein 3; U2 small nuclear RNA auxiliary factor 1-like protein 4; U2(RNU2) small nuclear RNA auxiliary factor 1-like 3; U2(RNU2) small nuclear RNA auxiliary factor 1-like protein 3; U2AF1-like 4; U2AF1-like protein 3; U2AF1L4 U2 small nuclear RNA auxiliary factor 1-like 4 FLJ35525 MGC33901 U2AF1-RS3 U2AF1L3 U2AF1L3V1 U2AF1RS3 U2af26 U2 small nuclear RNA auxiliary factor 1-like 3 U2(RNU2) small nuclear RNA auxiliary factor 1-like 3
Gene Aliases: U2AF1-RS3; U2AF1L3; U2AF1L3V1; U2AF1L4; U2AF1RS3; U2af26
UniProt ID: (Human) Q8WU68
Entrez Gene ID: (Human) 199746
Molecular Function:
RNA binding protein
nucleic acid binding
ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.