Invitrogen
Manufacturer Code:PA528510
Catalog # PIPA528510
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 23 and 227 of Human U2AF35 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FP793 RN RNU2AF1 U2AF35 U2AFBP U2 small nuclear RNA auxillary factor 1 U2 small nuclear ribonucleoprotein auxillary factor 35-KD subunit U2 snRNP auxiliary factor small subunit U2(RNU2) small nuclear RNA auxiliary factor binding protein splicing factor U2AF 35 kDa subunit splicing fac; Splicing factor U2AF 35 kDa subunit; Splicing factor U2AF 35 kDa subunit-like protein; splicing factor U2AF 35kDa subunit; U2 auxiliary factor 35 kDa subunit; U2 small nuclear ribonucleoprotein auxillary factor, 35-KD subunit; U2 small nuclear RNA auxiliary factor 1; U2 small nuclear RNA auxiliary factor 1-like protein 5; U2 small nuclear RNA auxillary factor 1; U2 snRNP auxiliary factor small subunit; U2(RNU2) small nuclear RNA auxiliary factor binding protein
Gene Aliases: FP793; RN; RNU2AF1; U2AF1; U2AF1L5; U2AF35; U2AFBP
UniProt ID: (Human) Q01081
Entrez Gene ID: (Human) 7307
Molecular Function:
RNA binding protein
nucleic acid binding
ribonucleoprotein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.