Novus Biologicals
Manufacturer Code:NBP180452
Catalog # NBP180452
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SNRPA. Peptide sequence MPGQMPPAQPLSENPPNHILFLTNLPEETNELMLSMLFNQFPGFKEVRLV. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Mud1 small nuclear ribonucleoprotein polypeptide A U1 snRNP A U1 snRNP-specific protein A U1-AU1 small nuclear ribonucleoprotein A U1AU1 small nuclear RNP-specific A; U1 small nuclear ribonucleoprotein A; U1 small nuclear RNP-specific A; U1 snRNP A; U1 snRNP-specific protein A
Gene Aliases: Mud1; SNRPA; U1-A; U1A
UniProt ID: (Human) P09012
Entrez Gene ID: (Human) 6626
Molecular Function: RNA binding protein mRNA processing factor mRNA splicing factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.