R&D Systems
Manufacturer Code:AF6236SP
Catalog # AF6236SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | Chinese hamster ovary cell line CHO-derived recominant human Tyrosylprotein Sulfotransferase 2/TPST2 Gln26-Ser377 (predicted) Accession # O60704 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.8.2.20 TPST-2 tyrosylprotein phosphotransferase 2 Tyrosylprotein Sulfotransferase 2 tyrosylprotein sulfotransferase 2protein-tyrosine sulfotransferase 2 tyrosylprotein sulfotransferase-2; Protein-tyrosine sulfotransferase 2; TPST-2; transport and golgi organization 13 homolog B; tyrosylprotein phosphotransferase 2; Tyrosylprotein sulfotransferase 2; tyrosylprotein sulfotransferase-2
Gene Aliases: TANGO13B; TPST2
UniProt ID: (Human) O60704
Entrez Gene ID: (Human) 8459
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.