Novus Biologicals
Manufacturer Code:NBP267797
Catalog # NBP267797
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | SD080-02 |
Immunogen | This Tyrosine Hydroxylase Antibody (SD080-02) is made against a recombinant protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: dystonia 14; DYT14 DYT5b EC 1.14.16 EC 1.14.16.2 TH TYH dystonia 14 Tyrosine 3-hydroxylase tyrosine 3-monooxygenase tyrosine hydroxylase; TH; Tyrosine 3-hydroxylase; Tyrosine 3-monooxygenase
Gene Aliases: DYT14; DYT5b; TH; TYH
UniProt ID: (Human) P07101
Entrez Gene ID: (Human) 7054
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.