Novus Biologicals
Manufacturer Code:NBP220770
Catalog # NBP220770
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human Tyrosinase. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CMM8 EC 1.14.18.1 LB24-AB Monophenol monooxygenase OCA1A OCAIA SHEP3 SK29-AB Tumor rejection antigen AB tyrosinase tyrosinase (oculocutaneous albinism IA); LB24-AB; Monophenol monooxygenase; oculocutaneous albinism IA; SK29-AB; Tumor rejection antigen AB; Tyrosinase
Gene Aliases: ATN; CMM8; OCA1; OCA1A; OCAIA; SHEP3; TYR
UniProt ID: (Human) Q15680
Entrez Gene ID: (Human) 7299
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.