Novus Biologicals
Manufacturer Code:NBP237364SS
Catalog # NBP237364SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 10E4E6 |
Immunogen | Purified recombinant fragment of human TWIST1 (AA: 9-74) expressed in E. Coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: acrocephalosyndactyly 3 ACS3 B-HLH DNA binding protein BHLHA38 bHLHa38BPES3 Class A basic helix-loop-helix protein 38 CRS1 H-twistblepharophimosis epicanthus inversus and ptosis 3 SCSBPES2 TWIST twist homolog 1 (Drosophila) TWIST homolog of drosophila twist-related protein 1; B-HLH DNA binding protein; bHLHa38; Class A basic helix-loop-helix protein 38; H-twist; twist basic helix-loop-helix transcription factor 1; twist homolog 1; TWIST homolog of drosophila; Twist-related protein 1
Gene Aliases: ACS3; BHLHA38; BPES2; BPES3; CRS; CRS1; CSO; SCS; TWIST; TWIST1
UniProt ID: (Human) Q15672
Entrez Gene ID: (Human) 7291
Molecular Function:
basic helix-loop-helix transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.