R&D Systems
Manufacturer Code:AF6230SP
Catalog # AF6230SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant human Twist-1 Met1-His202 Accession # Q15672 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: acrocephalosyndactyly 3 ACS3 B-HLH DNA binding protein BHLHA38 bHLHa38BPES3 BPES3 Class A basic helix-loop-helix protein 38 CRS1 H-twist H-twistblepharophimosis epicanthus inversus and ptosis 3 SCSBPES2 TWIST twist homolog 1 (Drosophila) TWIST homolog of drosophila Twist1 twist-related protein 1; B-HLH DNA binding protein; bHLHa38; Class A basic helix-loop-helix protein 38; H-twist; twist basic helix-loop-helix transcription factor 1; twist homolog 1; TWIST homolog of drosophila; Twist-related protein 1
Gene Aliases: ACS3; BHLHA38; BPES2; BPES3; CRS; CRS1; CSO; SCS; TWIST; TWIST1
UniProt ID: (Human) Q15672
Entrez Gene ID: (Human) 7291
Molecular Function: basic helix-loop-helix transcription factor nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.