Abnova
Manufacturer Code:PAB15951
Catalog # 89-117-981
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of mouse Tusc5. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Tusc5.
For Research Use Only
Protein Aliases: C130069F04Rik; Dispanin subfamily B member 1; DSPB1; LOST1; Trafficking regulator of GLUT4 1; Tumor suppressor candidate 5 homolog
Gene Aliases: C130069F04Rik; DSPB1; Lost1; Trarg1; Tusc5
UniProt ID: (Mouse) Q8C838
Entrez Gene ID: (Mouse) 237858
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.