Novus Biologicals
Manufacturer Code:NBP29396620UL
Catalog # NB022498
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 130-230 of human TUB (NP_003311.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: rd5 tubby (mouse) homolog tubby homolog (mouse) tubby homologue tubby protein homolog; tubby homolog; tubby homologue; Tubby protein homolog
Gene Aliases: rd5; RDOB; TUB
UniProt ID: (Human) P50607
Entrez Gene ID: (Human) 7275
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.