Invitrogen
Manufacturer Code:PA531164
Catalog # PIPA531164
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 4 and 263 of Human Tub |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: rd5 tubby homologue tubby protein homolog; retinal degeneration 5; tubby homolog; tubby homologue; Tubby protein; Tubby protein homolog
Gene Aliases: Rd5; RDOB; TUB
UniProt ID: (Human) P50607, (Mouse) P50586, (Rat) O88808
Entrez Gene ID: (Human) 7275, (Mouse) 22141, (Rat) 25609
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.