Abnova
Manufacturer Code:PAB15654
Catalog # 89-117-721
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to mouse Tspo. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised aganist synthetic peptide of Tspo.
For Research Use Only
Protein Aliases: benzodiazepine receptor, peripheral; Bzrp; IBP; isoquinoline-binding protein; Mitochondrial benzodiazepine receptor; PBR; Peripheral-type benzodiazepine receptor; PKBS; Translocator protein
Gene Aliases: Bzrp; IBP; Mbr; PBR; Tspo; Tspo1
UniProt ID: (Mouse) Q541E3
Entrez Gene ID: (Mouse) 12257
Molecular Function: receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.