Novus Biologicals
Manufacturer Code:NBP220744
Catalog # NBP220744
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the center region of human Tryptophanyl tRNA synthetase. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 6.1.1 EC 6.1.1.2 GAMMA-2 hWRS IFI53 IFP53trpRS Interferon-induced protein 53 TrpRS tryptophan tRNA ligase 1 cytoplasmic Tryptophan--tRNA ligase tryptophanyl-tRNA synthetase tryptophanyl-tRNA synthetase cytoplasmic WRS; hWRS; IFP53; Interferon-induced protein 53; T1-TrpRS; T2-TrpRS; TrpRS; tryptophan tRNA ligase 1, cytoplasmic; Tryptophan--tRNA ligase, cytoplasmic; Tryptophanyl-tRNA synthetase
Gene Aliases: GAMMA-2; IFI53; IFP53; WARS; WARS1; WRS
UniProt ID: (Human) P23381
Entrez Gene ID: (Human) 7453
Molecular Function: aminoacyl-tRNA synthetase ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.