Novus Biologicals
Manufacturer Code:NBP29452020UL
Catalog # NB023565
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 30-100 of human WRB (NP_004618.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Congenital heart disease 5 protein; congenital heart disease 5 protein10tryptophan-rich protein FLJ51808 tryptophan rich basic protein; Guided entry of tail-anchored proteins factor 1; Tail-anchored protein insertion receptor WRB; Tryptophan-rich basic protein
Gene Aliases: CHD5; GET1; WRB
UniProt ID: (Human) O00258
Entrez Gene ID: (Human) 7485
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.