Novus Biologicals
Manufacturer Code:NBP177559
Catalog # NBP177559
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide made to an N-terminal portion of the human Tryptophan hydroxylase 2 protein (between residues 100-200) [UniProt# Q8IWU9] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADHD7 EC 1.14.16 EC 1.14.16.4 FLJ37295 MGC138872 Neuronal tryptophan hydroxylase NTPHMGC138871 tryptophan 5-hydroxylase 2 Tryptophan 5-monooxygenase 2 tryptophan hydroxylase 2; Neuronal tryptophan hydroxylase; Tryptophan 5-hydroxylase 2; Tryptophan 5-monooxygenase 2
Gene Aliases: ADHD7; NTPH; TPH2
UniProt ID: (Human) Q8IWU9
Entrez Gene ID: (Human) 121278
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.