Invitrogen
Manufacturer Code:MA532981
Catalog # PIMA532981
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | WH-3 |
Immunogen | Recombit rabbit tryptophan hydroxylase. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: indoleacetic acid-5-hydroxylase; L-tryptophan hydroxylase; Neuronal tryptophan hydroxylase; Tryptophan 5-hydroxylase 1; Tryptophan 5-hydroxylase 2; Tryptophan 5-hydroxylase 2 1.14.16.4 Neuronal tryptophan hydroxylase Tryptophan 5-monooxygenase 2 Tph2 Ntph; Tryptophan 5-monooxygenase 1; Tryptophan 5-monooxygenase 2; tryptophan hydroxylase (tryptophan 5-monooxygenase)
Gene Aliases: ADHD7; NTPH; TPH; TPH1; TPH2; TPRH; TRPH
UniProt ID: (Human) P17752, (Rabbit) P17290, (Human) Q8IWU9, (Rat) P09810, (Rat) Q8CGU9
Entrez Gene ID: (Human) 7166, (Rabbit) 100009100, (Human) 121278, (Rabbit) 100344397, (Rat) 24848, (Rat) 317675
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.