Novus Biologicals
Manufacturer Code:NB300307
Catalog # NB300307
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser260 conjugated to KLH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 1.14.16 EC 1.14.16.4 indoleacetic acid-5-hydroxylase L-tryptophan hydroxylase MGC119994 TPH TPRH TRPH tryptophan 5-hydroxylase 1 Tryptophan 5-monooxygenase 1 tryptophan hydroxylase (tryptophan 5-monooxygenase) tryptophan hydroxylase 1; indoleacetic acid-5-hydroxylase; L-tryptophan hydroxylase; Tryptophan 5-hydroxylase 1; Tryptophan 5-monooxygenase 1; tryptophan hydroxylase (tryptophan 5-monooxygenase)
Gene Aliases: TPH; TPH1; TPRH; TRPH
UniProt ID: (Human) P17752
Entrez Gene ID: (Human) 7166
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.