Invitrogen
Manufacturer Code:PA5116095
Catalog # PIPA5116095
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human TXNRD2 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA118373 ESTM573010 KIAA1652 Selenoprotein Z SelZ TGR thioredoxin reductase 2 thioredoxin reductase 2 mitochondrial thioredoxin reductase 3 thioredoxin reductase beta thioredoxin reductase TR3 TR TR beta Tr3 TR-beta TRXR2 Trxrd2 Txnrd2; Selenoprotein Z; SelZ; Thioredoxin reductase 2, mitochondrial; thioredoxin reductase 3; thioredoxin reductase beta; Thioredoxin reductase TR3; TR beta; TR-beta
Gene Aliases: AA118373; ESTM573010; KIAA1652; SELZ; TGR; TR; TR-BETA; TR3; TRXR2; Trxrd2; TXNRD2
UniProt ID: (Human) Q9NNW7, (Mouse) Q9JLT4, (Rat) Q9Z0J5
Entrez Gene ID: (Human) 10587, (Mouse) 26462, (Rat) 50551
Molecular Function: dehydrogenase oxidase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.