Abnova
Manufacturer Code:PAB26829
Catalog # 89-128-473
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to mouse Trpc6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Trpc6.
For Research Use Only
Protein Aliases: AV025995; Calcium entry channel; LLHWJM002; LLHWJM003; LLHWJM004; mtrp6; Short transient receptor potential channel 6; Transient receptor protein 6; TRP-6; TrpC6; Trrp6
Gene Aliases: AV025995; LLHWJM002; LLHWJM003; LLHWJM004; mtrp6; TRP-6; Trp6; Trpc6; Trrp6
UniProt ID: (Mouse) Q61143
Entrez Gene ID: (Mouse) 22068
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.