Abnova
Manufacturer Code:PAB8504
Catalog # 89-133-714
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to mouse Trpa1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Trpa1.
For Research Use Only
Protein Aliases: ANKTM1; Ankyrin-like with transmembrane domains protein 1; Transient receptor potential cation channel subfamily A member 1; Wasabi receptor
Gene Aliases: Anktm1; Trpa1; TRPA1b
UniProt ID: (Mouse) Q8BLA8
Entrez Gene ID: (Mouse) 277328
Molecular Function:
ion channel
transporter
voltage-gated ion channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.