Abnova
Manufacturer Code:PAB24691
Catalog # 89-126-368
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to mouse Trp63. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of Trp63.
For Research Use Only
Protein Aliases: AI462811; Ket; MGC115972; p51/p63; p63; p73L; TAp63; TP63; Transformation-related protein 63; Trp53rp1; Tumor protein 63; Tumor protein p73-like
Gene Aliases: AI462811; Ket; P51/P63; P63; P73l; Tp63; Tp73l; Trp53rp1; Trp63
UniProt ID: (Mouse) O88898
Entrez Gene ID: (Mouse) 22061
Molecular Function: transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.