Novus Biologicals
Manufacturer Code:NBP229729AF532
Catalog # NP229729A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2G3 |
Immunogen | Human cardiac Troponin T. |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: ANMtroponin T slow skeletal muscle FLJ98147 MGC104241 Slow skeletal muscle troponin T STNT TNTS TNTtroponin T1 skeletal slow troponin T type 1 (skeletal slow) troponin-T1 skeletal slow; Slow skeletal muscle troponin T; sTnT; TnTs; troponin T type 1 (skeletal, slow); Troponin T, slow skeletal muscle; troponin-T1, skeletal, slow
Gene Aliases: ANM; NEM5; STNT; TNNT1; TNT; TNTS
UniProt ID: (Human) P13805
Entrez Gene ID: (Human) 7138
Molecular Function: actin binding motor protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.