EMD Millipore
Manufacturer Code:MABT136525UL
Catalog # MABT136525
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 7F8B8 |
Immunogen | KLH-conjugated linear peptides corresponding to 24 amino acids from the N-terminal region of isoform 5 and 4 amino acids from the N-terminal region of isoform 2 of human tropomyosin-1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Stable for 1 year at 2-8°C from date of receipt. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: Tropomyosin alpha-1 chainAlpha-tropomyosinTropomyosin-1
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.