Novus Biologicals
Manufacturer Code:NBP189896
Catalog # NBP189896
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:KVIESRAQKDEEKMEIQEIQLKEAKHIAEDADRKYEEVARKLVIIESDLERAEERAELSEGQVRQLEEQLRIMDQTLKALMAAEDKYSQKEDRYEEEIKVLSDKLKEAETRAEFAE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha tropomyosin alpha-tropomyosin C15orf13 cardiomyopathy hypertrophic 3 chromosome 15 open reading frame 13 CMD1Y CMH3 HTM-alpha sarcomeric tropomyosin kappa TMSA tropomyosin 1 (alpha) tropomyosin 1 (alpha) isoform 1 tropomyosin 1 (alpha) isoform 2 tropomyosin 1 (alpha) isoform 3 tropomyosin 1 (alpha) isoform 4 tropomyosin 1 (alpha) isoform 5 tropomyosin 1 (alpha) isoform 6 tropomyosin 1 (alpha) isoform 7 tropomyosin alpha-1 chain Tropomyosin-1; Alpha-tropomyosin; cardiomyopathy, hypertrophic 3; epididymis secretory protein Li 265; sarcomeric tropomyosin kappa; Tropomyosin alpha-1 chain; Tropomyosin-1
Gene Aliases: C15orf13; CMD1Y; CMH3; HEL-S-265; HTM-alpha; LVNC9; TMSA; TPM1
UniProt ID: (Human) P09493
Entrez Gene ID: (Human) 7168
Molecular Function: actin binding motor protein actin family cytoskeletal protein cytoskeletal protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.