Novus Biologicals
Manufacturer Code:NBP147900
Catalog # NBP147900
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2B8 |
Immunogen | Full-length protein expressed in 293T cell transfected with human NTRK3 expression vector |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.10 EC 2.7.10.1 ETS related protein-neurotrophic receptor tyrosine kinase fusion protein ETV6-NTRK3 fusion gp145(trkC) GP145-TrkC Neurotrophic tyrosine kinase receptor type 3 neurotrophic tyrosine kinase receptor type 3 NT 3 receptor NT-3 growth factor receptor Trk-C TrkC tyrosine kinase TRKCneurotrophin 3 receptor tyrosine kinase receptor C; ETS related protein-neurotrophic receptor tyrosine kinase fusion protein; ETV6-NTRK3 fusion; GP145-TrkC; Neurotrophic tyrosine kinase receptor type 3; neurotrophic tyrosine kinase, receptor, type 3; NT-3 growth factor receptor; Trk-C; TrkC tyrosine kinase; tyrosine kinase receptor C
Gene Aliases: gp145(trkC); GP145-TrkC; NTRK3; TRKC
UniProt ID: (Human) Q16288
Entrez Gene ID: (Human) 4916
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.