Invitrogen
Manufacturer Code:OST00118G
Catalog # PIOST00118G
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the extracellular domain of mouse TrkB |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: BDNF/NT-3 growth factors receptor; GP145-TrkB/GP95-TrkB; neural receptor protein-tyrosine kinase (trkB); Neurotrophic tyrosine kinase receptor type 2; Neurotrophic tyrosine kinase receptor type 2 TrkB tyrosine kinase GP145-TrkB/GP95-TrkB Trk-B BDNF/NT-3 growth factors receptor; neurotrophic tyrosine receptor kinase type 2; Trk-B; TrkB tyrosine kinase
Gene Aliases: GP145-TrkB/GP95-TrkB; Ntrk2; RATTRKB1; Tkrb; trk-B; Trkb; TRKB1
UniProt ID: (Mouse) P15209, (Rat) Q63604
Entrez Gene ID: (Mouse) 18212, (Rat) 25054
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.