R&D Systems
Manufacturer Code:AF1494SP
Catalog # AF1494SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse TrkB Cys32-His429 Accession # P15209 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: BDNF-tropomyosine receptor kinase B; BDNF/NT-3 growth factors receptor; BDNF/NT-3 growth factors receptor EC 2.7.10 EC 2.7.10.1 GP145-TrkB Neurotrophic tyrosine kinase receptor type 2 neurotrophic tyrosine kinase receptor type 2 NTRK2 trk-B TrkB tyrosine kinase TRKBBDNF-tropomyosine receptor kinase B tyrosine kinase receptor B; GP145-TrkB; Neurotrophic tyrosine kinase receptor type 2; neurotrophic tyrosine kinase, receptor, type 2; Trk-B; TrkB tyrosine kinase; Tropomyosin-related kinase B; tyrosine kinase receptor B
Gene Aliases: GP145-TrkB; NTRK2; trk-B; TRKB
UniProt ID: (Human) Q16620
Entrez Gene ID: (Human) 4915
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.