R&D Systems
Manufacturer Code:MAB1752100
Catalog # MAB1752100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Neutralization (Neu) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 994307 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human TrkA Ala33-Glu407 Accession No. P04629 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70°C as supplied. 1 month 2 to 8°C under sterile conditions after reconstitution. 6 months -20 to -70°C under sterile conditions a |
Mouse Monoclonal Antibody
Protein Aliases: DKFZp781I14186 EC 2.7.10 EC 2.7.10.1 MTChigh affinity nerve growth factor receptor Neurotrophic tyrosine kinase receptor type 1 neurotrophic tyrosine kinase receptor type 1 NTRK1 NTRK-1 p140-TrkA TRK1-transforming tyrosine kinase protein Trk-A TRKAOncogene TRK TRKTRK1 tyrosine kinase receptor A; gp140trk; High affinity nerve growth factor receptor; Neurotrophic tyrosine kinase receptor type 1; neurotrophic tyrosine kinase, receptor, type 1; Oncogene TRK; p140-TrkA; Trk-A; TRK1-transforming tyrosine kinase protein; Tropomyosin-related kinase A; Tyrosine kinase receptor; Tyrosine kinase receptor A
Gene Aliases: MTC; NTRK1; p140-TrkA; TRK; Trk-A; TRK1; TRKA
UniProt ID: (Human) P04629
Entrez Gene ID: (Human) 4914
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.