Invitrogen
Manufacturer Code:OST00115W
Catalog # PIOST00115W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the extracellular domain of rat TrkA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: High affinity nerve growth factor receptor; High affinity nerve growth factor receptor NTRK1 p140-TrkA Trk-A Neurotrophic tyrosine kinase receptor type 1 Slow nerve growth factor receptor; Neurotrophic tyrosine kinase receptor type 1; p140-TrkA; Slow nerve growth factor receptor; trk; Trk-A; TrkA neurotrophin receptor; trkA proto-oncogene receptor; tropomyosin-related kinase A
Gene Aliases: Ntrk1; Trk; Trka
UniProt ID: (Rat) P35739
Entrez Gene ID: (Rat) 59109
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.