Invitrogen
Manufacturer Code:PA538053
Catalog # PIPA538053
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | 19 amino acid peptide near the carboxy terminus of human TICRR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: SLD3 TICRR C15orf42 TopBP1-interacting checkpoint and replication regulator; TOPBP1-interacting checkpoint and replication regulator; topBP1-interacting replication-stimulating protein; TopBP1-interacting, replication-stimulating protein; Treslin
Gene Aliases: C15orf42; SLD3; TICRR; Treslin
UniProt ID: (Human) Q7Z2Z1
Entrez Gene ID: (Human) 90381
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.