Invitrogen
Manufacturer Code:OSR00084W
Catalog # PIOSR00084W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Fruit fly |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide from Drosophila melanogaster gustatory receptor trehalose 1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CG15779-PA; Drosophila gustatory receptor trehalose 1 Tre1; Gr5a-PA; Gustatory receptor 5a for trehalose; gustatory receptor LU.7; Trehalose receptor; trehalose sensitivity; Trehalose-sensitivity
Gene Aliases: CG15779; Dmel5a; Dmel\CG15779; Dmel_CG15779; Gr5; GR5A; Gr5A1; GRLU.7; GrLU7; han; LU.7; Tre
UniProt ID: (Fruit fly) Q9W497
Entrez Gene ID: (Fruit fly) 44873
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.