Novus Biologicals
Manufacturer Code:NBP267226
Catalog # NBP267226
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JF1016 |
Immunogen | Recombinant Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: ATTR; ATTR HsT2651 PALBTBPA Prealbumin prealbumin amyloidosis type I thyroxine-binding prealbumin transthyretin; carpal tunnel syndrome 1; epididymis luminal protein 111; Prealbumin; prealbumin, amyloidosis type I; TBPA; thyroxine-binding prealbumin; Transthyretin
Gene Aliases: CTS; CTS1; HEL111; HsT2651; PALB; TBPA; TTR
UniProt ID: (Human) P02766
Entrez Gene ID: (Human) 7276
Molecular Function:
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.