Invitrogen
Manufacturer Code:PA580197
Catalog # PIPA580197
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E.coli-derived rat Prealbumin recombit protein (Position: G21-N147). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Prealbumin; prealbumin, amyloidosis type I; TBPA; Transthyretin; Transthyretin Prealbumin TBPA Ttr Tt
Gene Aliases: AA408768; AI787086; D17860; Lr1; prealbumin; Tbpa; Tt; Ttr
UniProt ID: (Mouse) P07309, (Rat) P02767
Entrez Gene ID: (Mouse) 22139, (Rat) 24856
Molecular Function: transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.