Invitrogen
Manufacturer Code:PA520742
Catalog # PIPA520742
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Chicken |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 17 amino acid peptide near the center of human Transthyretin. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Chicken Polyclonal Antibody
For Research Use Only
Protein Aliases: ATTR; carpal tunnel syndrome 1; CTS CTS1 HsT2651 PALB TBPA ATTR carpal tunnel syndrome 1 prealbumin amyloidosis type I thyroxine-binding prealbumin; epididymis luminal protein 111; Prealbumin; prealbumin, amyloidosis type I; TBPA; thyroxine-binding prealbumin; Transthyretin
Gene Aliases: AA408768; AI787086; CTS; CTS1; D17860; HEL111; HsT2651; Lr1; PALB; prealbumin; TBPA; Tt; TTR
UniProt ID: (Human) P02766, (Mouse) P07309, (Rat) P02767
Entrez Gene ID: (Human) 7276, (Mouse) 22139, (Rat) 24856
Molecular Function:
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.