Novus Biologicals
Manufacturer Code:NBP29476520UL
Catalog # NB024047
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 800 to the C-terminus of human TNPO1 (NP_002261.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: importin 2; importin beta 2; importin beta 2 Importin beta-2 IPO2 karyopherin (importin) beta 2 MIP1Karyopherin beta-2 MIPM9 region interaction protein transportin 1 transportin-1 TRNKPNB2importin 2; Importin beta-2; karyopherin (importin) beta 2; Karyopherin beta-2; M9 region interaction protein; MIP; Transportin-1
Gene Aliases: IPO2; KPNB2; MIP; MIP1; TNPO1; TRN
UniProt ID: (Human) Q92973
Entrez Gene ID: (Human) 3842
Molecular Function:
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.