Invitrogen
Manufacturer Code:PA543192
Catalog # PIPA543192
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human TKT |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: epididymis luminal protein 107; epididymis secretory protein Li 48; P68; TK; Transketolase; transketolase (Wernicke-Korsakoff syndrome)
Gene Aliases: HEL-S-48; HEL107; p68; TK; TKT; TKT1
UniProt ID: (Bovine) Q6B855, (Human) P29401, (Mouse) P40142, (Rat) P50137
Entrez Gene ID: (Bovine) 445425, (Human) 7086, (Pig) 100127150, (Rabbit) 100350406, (Mouse) 21881, (Rat) 64524
Molecular Function:
dehydrogenase
lyase
oxidoreductase
transferase
transketolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.