Invitrogen
Manufacturer Code:MA517922
Catalog # PIMA517922
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | R17217.1.4 |
Immunogen | Mouse CD71 |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: CD71; CD71 T9 TFR TFR1 TR TRFR p90 transferrin receptor protein 1; mammary tumor virus receptor 1; TR; Transferrin receptor protein 1
Gene Aliases: 2610028K12Rik; AI195355; AI426448; AU015758; CD71; E430033M20Rik; Mtvr-1; Mtvr1; p90; TFR; TFR1; Tfrc; TR; Trfr
UniProt ID: (Mouse) Q62351
Entrez Gene ID: (Mouse) 22042
Molecular Function:
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.