Proteintech
Manufacturer Code:661711IG150UL
Catalog # 50-173-6605
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B4C6 |
Immunogen | Transferrin Fusion Protein Ag11668 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Beta 1 metal binding globulin PRO1557 PRO2086 Serotransferrin Siderophilin TF Transferrin; Beta-1 metal-binding globulin; epididymis secretory sperm binding protein Li 71p; Serotransferrin; Siderophilin; Transferrin
Gene Aliases: HEL-S-71p; PRO1400; PRO1557; PRO2086; TF; TFQTL1
UniProt ID: (Human) P02787
Entrez Gene ID: (Human) 7018
Molecular Function:
hydrolase
protease
serine protease
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.