R&D Systems
Manufacturer Code:AF3987
Catalog # AF3987
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse plasma-derived Transferrin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-1 metal-binding globulin; Beta-1 metal-binding globulin DKFZp781D0156 EC 3.4.21 PRO1557 PRO2086 serotransferrin Siderophilin TF transferrin; epididymis secretory sperm binding protein Li 71p; Serotransferrin; Siderophilin; Transferrin
Gene Aliases: HEL-S-71p; PRO1400; PRO1557; PRO2086; TF; TFQTL1
UniProt ID: (Human) P02787
Entrez Gene ID: (Human) 7018
Molecular Function: hydrolase protease serine protease transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.