R&D Systems
Manufacturer Code:MAB7895SP
Catalog # MAB7895SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 874420 |
Immunogen | Chinese hamster ovary cell line CHO-derived recombinant human Transcobalamin II Glu19-Trp427 Accession # P20062 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: D22S676 D22S750 macrocytic anemia TC TC-2 TC2II TCII TCN2 transcobalamin II macrocytic anemia transcobalamin IITC II transcobalamin-2 vitamin B12-binding protein 2; macrocytic anemia; TC II; TC-2; Transcobalamin II; transcobalamin II; macrocytic anemia; Transcobalamin-2; vitamin B12-binding protein 2
Gene Aliases: D22S676; D22S750; II; TC; TC II; TC-2; TC2; TCII; TCN2
UniProt ID: (Human) P20062
Entrez Gene ID: (Human) 6948
Molecular Function: cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.