Novus Biologicals
Manufacturer Code:NBP298341B
Catalog # NB082043
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with purified recombinant Transcobalamin II (O88968 Met 1-Trp 430). |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: D22S676 D22S750 macrocytic anemia TC TC-2 TC2II TCII transcobalamin II macrocytic anemia transcobalamin IITC II transcobalamin-2 vitamin B12-binding protein 2; macrocytic anemia; TC II; TC-2; Transcobalamin II; transcobalamin II; macrocytic anemia; Transcobalamin-2; vitamin B12-binding protein 2
Gene Aliases: D22S676; D22S750; II; TC; TC II; TC-2; TC2; TCII; TCN2
UniProt ID: (Human) P20062
Entrez Gene ID: (Human) 6948
Molecular Function: cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.