Novus Biologicals
Manufacturer Code:NBP132121
Catalog # NBP132121
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human Transaldolase. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dihydroxyacetone transferase; dihydroxyacetone transferase EC 2.2.1.2 glycerone transferase TAL TALDO TALDOR TALH TAL-H transaldolase transaldolase 1; glycerone transferase; testicular secretory protein Li 56; Transaldolase
Gene Aliases: TAL; TAL-H; TALDO; TALDO1; TALDOR; TALH
UniProt ID: (Human) P37837
Entrez Gene ID: (Human) 6888
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.