Proteintech
Manufacturer Code:102961AP150UL
Catalog # 50-172-6187
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Torsin A Fusion Protein Ag0358 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DQ2 Dystonia 1 protein DYT1 TOR1A Torsin 1A Torsin A Torsin family 1 member A TorsinA; Dystonia 1 protein; dystonia 1, torsion (autosomal dominant; dystonia 1, torsion (autosomal dominant; torsin A); torsin A; Torsin ATPase 1; Torsin ATPase-1A; Torsin family 1 member A; torsin family 1, member A (torsin A); Torsin-1A
Gene Aliases: DQ2; DYT1; TA; TOR1A; TORA; torsinA
UniProt ID: (Human) O14656, (Rat) Q68G38, (Mouse) Q9ER39
Entrez Gene ID: (Human) 1861, (Rat) 266606, (Mouse) 30931
Molecular Function: chaperone
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.