Novus Biologicals
Manufacturer Code:NBP130482AF700
Catalog # NB130482AFB
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen for this product maps to a region between residue 715 and 765 of human DNA topoisomerase I using the numbering given in entry NP_003277.1 (GeneID 7150). |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DNA topoisomerase 1; DNA topoisomerase 1 DNA topoisomerase I EC 5.99.1.2 TOPI topoisomerase (DNA) I type I DNA topoisomerase; DNA topoisomerase I; type I DNA topoisomerase
Gene Aliases: TOP1; TOPI
UniProt ID: (Human) P11387
Entrez Gene ID: (Human) 7150
Molecular Function:
DNA binding protein
DNA topoisomerase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.