Novus Biologicals
Manufacturer Code:NB10092303
Catalog # NB10092303
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide,corresponding to amino acids 51-100 of Human Topo IIIbeta-1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
For Research Use Only
Protein Aliases: DNA topoisomerase 3-beta-1; DNA topoisomerase III beta-1; topoisomerase III beta
Gene Aliases: TOP3B; TOP3B1
UniProt ID: (Human) O95985
Entrez Gene ID: (Human) 8940
Molecular Function:
enzyme modulator
isomerase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.