Proteintech
Manufacturer Code:112781AP150UL
Catalog # 50-172-7024
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Tom22 Fusion Protein Ag1797 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1C9 2 hTom22 MST065 MSTP065 TOM22 TOMM22; 1C9-2; hTom22; Mitochondrial import receptor subunit TOM22 homolog; mitochondrial import receptor Tom22; rTOM22; Translocase of outer membrane 22 kDa subunit homolog; translocase of outer mitochondrial membrane 22 homolog
Gene Aliases: 1C9-2; 2310047D01; MST065; MSTP065; rTOM22; TOM22; TOMM22
UniProt ID: (Human) Q9NS69, (Rat) Q75Q41, (Mouse) Q9CPQ3
Entrez Gene ID: (Human) 56993, (Rat) 300075, (Mouse) 223696
Molecular Function:
receptor
transfer/carrier protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.