R&D Systems
Manufacturer Code:MAB7630100
Catalog # MAB7630100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 805125 |
Immunogen | <em>E.coli</em>-derived recombinant human Tmp21/p23 Ile32-Arg185 Accession # P49755 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 21 kDa transmembrane trafficking protein; 21 kDa transmembrane-trafficking protein; 21 kDa transmembrane-trafficking protein p23 P24(DELTA) p24delta S31I125 S31III125 TMED10 TMP21Tmp-21-I transmembrane emp24 domain-containing protein 10 transmembrane emp24-like trafficking protein 10 (yeast) Transmembrane protein Tmp2121 kDa transmembrane trafficking protein; p23; p24 family protein delta-1; p24delta; p24delta1; Protein TMED10; S31I125; S31III125; testicular tissue protein Li 206; Tmp-21-I; Transmembrane emp24 domain-containing protein 10; transmembrane emp24-like trafficking protein 10; Transmembrane protein Tmp21
Gene Aliases: p23; P24(DELTA); p24d1; S31I125; S31III125; TMED10; Tmp-21-I; TMP21
UniProt ID: (Human) P49755
Entrez Gene ID: (Human) 10972
Molecular Function:
membrane traffic protein
transfer/carrier protein
vesicle coat protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.